AMBH2D6FDD03 Ambeed, Inc.

(2-(Methylamino)phenyl)methanol

  • MDL number MFCD09031020
  • Cas Number : 29055-08-1
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 149
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form Light yellow to yellow powder or crystals or liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 284.1�t 760 mmHg
Product Link https://www.ambeed.com/products/29055-08-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/29055-08-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart