AMBH2D6FDD0B Ambeed, Inc.

(2-Nitrophenyl)(phenyl)methanol

  • MDL number MFCD09030980
  • Cas Number : 5176-12-5
  • Molecular Weight : 229.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/5176-12-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/5176-12-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart