AMBH2D6FDD16 Ambeed, Inc.

2-(Pyridin-2-yloxy)acetic acid

  • MDL number MFCD01646171
  • Cas Number : 58530-50-0
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/58530-50-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/58530-50-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart