AMBH2D6FDEB3 Ambeed, Inc.

1-Boc-(3S,4S)-3-amino-4-hydroxypyrrolidine

  • MDL number MFCD11111139
  • Cas Number : 190792-74-6
  • Molecular Weight : 202.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-144
Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 308.8�t 760 mmHg
Product Link https://www.ambeed.com/products/190792-74-6.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/190792-74-6.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart