AMBH2D6FDF2B Ambeed, Inc.

tert-Butyl (5-aminobenzo[d]isoxazol-3-yl)carbamate

  • MDL number MFCD09834801
  • Cas Number : 380629-73-2
  • Molecular Weight : 249.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/380629-73-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/380629-73-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart