AMBH2D6FDFA7 Ambeed, Inc.

Potassium trifluoro(3-(methylsulfonyl)phenyl)borate

  • MDL number MFCD28143971
  • Cas Number : 1985700-16-0
  • Molecular Weight : 262.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1985700-16-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1985700-16-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart