AMBH2D6FDFE4 Ambeed, Inc.

(R)-1-(3,4-Dimethoxyphenyl)ethanamine

  • MDL number MFCD06761897
  • Cas Number : 100570-24-9
  • Molecular Weight : 181.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 128.2°C
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 272°C at 760 mmHg
Product Link https://www.ambeed.com/products/100570-24-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/100570-24-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart