AMBH2D6FE01C Ambeed, Inc.

2-Chloro-6-(trifluoromethoxy)phenol

  • MDL number MFCD18393866
  • Cas Number : 1036713-42-4
  • Molecular Weight : 212.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1036713-42-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1036713-42-4.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart