AMBH2D6FE030 Ambeed, Inc.

(R)-2-Phenylpyrrolidine

  • MDL number MFCD06762513
  • Cas Number : 56523-47-8
  • Molecular Weight : 147.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 98.3°C
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 237°C at 760 mmHg
Product Link https://www.ambeed.com/products/56523-47-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/56523-47-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart