AMBH2D6FE040 Ambeed, Inc.

2-Chloro-6-methoxyisonicotinonitrile

  • MDL number MFCD18204709
  • Cas Number : 1256788-36-9
  • Molecular Weight : 168.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1256788-36-9.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/1256788-36-9.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart