AMBH2D6FE196 Ambeed, Inc.

(4-(9-Phenyl-9H-carbazol-3-yl)phenyl)boronic acid

  • MDL number MFCD30063200
  • Cas Number : 1240963-55-6
  • Molecular Weight : 363.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1240963-55-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1240963-55-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart