AMBH2D6FEEA5 Ambeed, Inc.

6-Bromo-2-(trifluoromethyl)pyrimidin-4-amine

  • MDL number MFCD22371631
  • Cas Number : 1378259-23-4
  • Molecular Weight : 242.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1378259-23-4.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/1378259-23-4.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart