AMBH2D6FEEAA Ambeed, Inc.

6-Fluoro-1-methyl-1H-indole-2-carboxylic acid

  • MDL number MFCD07364602
  • Cas Number : 893731-12-9
  • Molecular Weight : 193.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/893731-12-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/893731-12-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart