AMBH2D6FEECA Ambeed, Inc.

1-(tert-Butoxycarbonyl)-4-oxopyrrolidine-2-carboxylic acid

  • MDL number MFCD02668169
  • Cas Number : 876317-19-0
  • Molecular Weight : 229.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155
Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/876317-19-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/876317-19-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart