AMBH2D6FEF2C Ambeed, Inc.

Benzo[d][1,2,3]thiadiazole

  • MDL number MFCD00047265
  • Cas Number : 273-77-8
  • Molecular Weight : 136.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 90.8
Storage temp. Sealed in dry,Room Temperature
Physical Form Red-brown to brown sticky oil to solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 220.5�t 760 mmHg
Product Link https://www.ambeed.com/products/273-77-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/273-77-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart