AMBH2D6FEF38 Ambeed, Inc.

Sodium 1,2,4-triazol-1-ide

  • MDL number MFCD00044437
  • Cas Number : 41253-21-8
  • Molecular Weight : 91.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 295°C
Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/41253-21-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/41253-21-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart