AMBH2D6FEF70 Ambeed, Inc.

2-Iodo-1,3,4-thiadiazole

  • MDL number MFCD11112108
  • Cas Number : 332133-91-2
  • Molecular Weight : 212.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/332133-91-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/332133-91-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart