AMBH2D6FEFC3 Ambeed, Inc.

1,3,5-Trimethylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD00154763
  • Cas Number : 4401-71-2
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/4401-71-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/4401-71-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart