AMBH2D6FF035 Ambeed, Inc.

1,8-Naphthyridin-2-amine

  • MDL number MFCD01250851
  • Cas Number : 15992-83-3
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 322.6�t 760 mmHg
Iupac Name [1,8]naphthyridin-2-amine
Inchi Code 1S/C8H7N3/c9-7-4-3-6-2-1-5-10-8(6)11-7/h1-5H,(H2,9,10,11)
Inchi Key CRADWWWVIYEAFR-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 322.6�t 760 mmHg
Product Link https://www.ambeed.com/products/15992-83-3.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/15992-83-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart