AMBH2D6FF076 Ambeed, Inc.

4'-Decyl-[1,1'-biphenyl]-4-carbonitrile

  • MDL number MFCD01218035
  • Cas Number : 59454-35-2
  • Molecular Weight : 319.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/59454-35-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/59454-35-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart