AMBH2D6FF09B Ambeed, Inc.

(S)-6-Acetamido-2-(2-((S)-2-acetamido-4-methylpentanamido)acetamido)-N-(4-methyl-2-oxo-2H-chromen-7-yl)hexanamide

  • MDL number MFCD22380596
  • Cas Number : 660847-06-3
  • Molecular Weight : 557.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/660847-06-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/660847-06-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart