AMBH2D6FF0B0 Ambeed, Inc.

4-Methoxy-2-methyl-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD12924602
  • Cas Number : 307951-52-6
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/307951-52-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/307951-52-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart