AMBH2D6FF12A Ambeed, Inc.

4-(4-(Trifluoromethyl)phenyl)-1H-imidazole

  • MDL number MFCD08669147
  • Cas Number : 37148-86-0
  • Molecular Weight : 212.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/37148-86-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/37148-86-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart