AMBH2D6FF19A Ambeed, Inc.

1-Chloro-2,4-dimethyl-5-nitrobenzene

  • MDL number MFCD00547853
  • Cas Number : 69383-68-2
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-43°C
Purity 95
Flash Point 117.5
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 270.8°C at 760 mmHg
Product Link https://www.ambeed.com/products/69383-68-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/69383-68-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart