AMBH2D6FF1B6 Ambeed, Inc.

1-Cyclopropyl-1-phenylethanol

  • MDL number MFCD00019243
  • Cas Number : 5558-04-3
  • Molecular Weight : 162.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/5558-04-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/5558-04-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart