AMBH2D6FF1BE Ambeed, Inc.

1-Ethyl-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD00956664
  • Cas Number : 25939-81-5
  • Molecular Weight : 161.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 257.4�t 760 mmHg
Product Link https://www.ambeed.com/products/25939-81-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/25939-81-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart