AMBH2D6FF1D8 Ambeed, Inc.

1-Fluoro-4-nitronaphthalene

  • MDL number MFCD00156398
  • Cas Number : 341-92-4
  • Molecular Weight : 191.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 144.1
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 314.6�t 760 mmHg
Product Link https://www.ambeed.com/products/341-92-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/341-92-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart