AMBH2D6FF1E3 Ambeed, Inc.

TAS-103 dihydrochloride

  • MDL number MFCD00943251
  • Cas Number : 174634-09-4
  • Molecular Weight : 369.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-((2-(Dimethylamino)ethyl)amino)-3-hydroxy-7H-indeno[2,1-c]quinolin-7-one hydrochloride
Purity 98
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/174634-09-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/174634-09-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart