AMBH2D6FF1E6 Ambeed, Inc.

tert-Butyl 3-propoxypyrrolidine-1-carboxylate

  • MDL number MFCD08533291
  • Cas Number : 1175835-99-0
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1175835-99-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1175835-99-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart