AMBH2D6FF1F6 Ambeed, Inc.

Ethyl 2-amino-2-(3-chloropyridin-4-yl)acetate hydrochloride

  • MDL number MFCD30186146
  • Cas Number : 1956319-58-6
  • Molecular Weight : 251.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1956319-58-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1956319-58-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart