AMBH2D6FF1FD Ambeed, Inc.

6-Chloro-7,8-dimethoxy-1-(4-methoxyphenyl)-2,3,4,5-tetrahydro-1H-benzo[d]azepine

  • MDL number MFCD08460175
  • Cas Number : 67287-53-0
  • Molecular Weight : 347.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-145
Purity 95
Flash Point 243.6
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 479.2�t 760 mmHg
Product Link https://www.ambeed.com/products/67287-53-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/67287-53-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart