AMBH2D6FF209 Ambeed, Inc.

1H-Benzo[d]imidazol-7-amine

  • MDL number MFCD09054675
  • Cas Number : 4331-29-7
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 273.2
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 476�t 760 mmHg
Product Link https://www.ambeed.com/products/4331-29-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/4331-29-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart