AMBH2D6FF25D Ambeed, Inc.

AZD1152-HQPA

  • MDL number MFCD17392583
  • Cas Number : 722544-51-6
  • Molecular Weight : 507.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/722544-51-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/722544-51-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart