AMBH2D6FF5E4 Ambeed, Inc.

1-((4-Fluorophenyl)sulfonyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD12028498
  • Cas Number : 1171916-98-5
  • Molecular Weight : 273.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1171916-98-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1171916-98-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart