AMBH2D6FF665 Ambeed, Inc.

Ethyl 2-piperazin-1-yl-thiazole-4-carboxylate

  • MDL number MFCD09910379
  • Cas Number : 104481-24-5
  • Molecular Weight : 241.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/104481-24-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/104481-24-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart