AMBH2D6FF68C Ambeed, Inc.

2-(2-Methyl-1H-imidazol-1-yl)benzoic acid

  • MDL number MFCD09065012
  • Cas Number : 159589-71-6
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232.1
Purity 97
Flash Point 213.3
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 429.1�t 760 mmHg
Product Link https://www.ambeed.com/products/159589-71-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/159589-71-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart