AMBH2D6FF7FE Ambeed, Inc.

2-Acetyl-3,5(6)-dimethylpyrazine, mixture of isomers

  • MDL number MFCD29036871
  • Cas Number : 72797-17-2
  • Molecular Weight : 300.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/72797-17-2.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/72797-17-2.html
pictograms GHS07
hstatements H227-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart