AMBH2D6FF81B Ambeed, Inc.

PD1-PDL1 inhibitor 2

  • MDL number MFCD29905455
  • Cas Number : 1675203-84-5
  • Molecular Weight : 419.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-(2-(((2-Methoxy-6-((2-methyl-[1,1'-biphenyl]-3-yl)methoxy)pyridin-3-yl)methyl)amino)ethyl)acetamide
Purity 98
Flash Point 323.6
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 611.4�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1675203-84-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1675203-84-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart