AMBH2D6FF865 Ambeed, Inc.

Diethyl 1-benzyl-4-phenyl-1,4-dihydropyridine-3,5-dicarboxylate

  • MDL number MFCD20267716
  • Cas Number : 120533-76-8
  • Molecular Weight : 391.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/120533-76-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/120533-76-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart