AMBH2D6FF90B Ambeed, Inc.

LY2606368 dihydrochloride

  • MDL number N/A
  • Cas Number : 1234015-54-3
  • Molecular Weight : 438.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1234015-54-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1234015-54-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart