AMBH2D6FF92C Ambeed, Inc.

SH-4-54

  • MDL number MFCD28143917
  • Cas Number : 1456632-40-8
  • Molecular Weight : 610.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(N-(4-cyclohexylbenzyl)-2-(2,3,4,5,6-pentafluoro-N-methylphenylsulfonamido)acetamido)benzoic acid
Purity 98
Flash Point 387.5
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 717.2�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1456632-40-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1456632-40-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart