AMBH2D6FF994 Ambeed, Inc.

Vidofludimus

  • MDL number MFCD18633262
  • Cas Number : 717824-30-1
  • Molecular Weight : 355.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-((3-Fluoro-3'-methoxy-[1,1'-biphenyl]-4-yl)carbamoyl)cyclopent-1-enecarboxylic acid
Purity 98
Flash Point 297
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 567.478�t 760 mmHg
Product Link https://www.ambeed.com/products/717824-30-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/717824-30-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart