AMBH2D6FF9B4 Ambeed, Inc.

tert-Butyl 4-(1-amino-2-hydroxyethyl)piperidine-1-carboxylate hydrochloride

  • MDL number MFCD30489019
  • Cas Number : 2044704-67-6
  • Molecular Weight : 280.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2044704-67-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2044704-67-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart