AMBH2D6FFA6F Ambeed, Inc.

Bicyclo[4.2.0]octa-1,3,5-trien-7-ylmethanol

  • MDL number MFCD09752655
  • Cas Number : 15100-35-3
  • Molecular Weight : 134.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/15100-35-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/15100-35-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart