AMBH2D6FFB02 Ambeed, Inc.

6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-6-carboxylic acid hydrochloride

  • MDL number MFCD30181971
  • Cas Number : 1965310-16-0
  • Molecular Weight : 188.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1965310-16-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1965310-16-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart