AMBH2D6FFB04 Ambeed, Inc.

Mono-O-AcetylGanciclovir

  • MDL number MFCD09840659
  • Cas Number : 88110-89-8
  • Molecular Weight : 297.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/88110-89-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/88110-89-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart