AMBH2D6FFB17 Ambeed, Inc.

Benzo[c][1,2,5]thiadiazol-5-amine

  • MDL number MFCD00185826
  • Cas Number : 874-37-3
  • Molecular Weight : 151.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-115
Purity 97
Flash Point 137.1
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 303�t 760 mmHg
Product Link https://www.ambeed.com/products/874-37-3.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/874-37-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart