AMBH2D7002BA Ambeed, Inc.

2,4,6-Trichloro-5-cyanopyrimidine

  • MDL number MFCD07636745
  • Cas Number : 3029-64-9
  • Molecular Weight : 208.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 320.8�t 760 mmHg
Product Link https://www.ambeed.com/products/3029-64-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/3029-64-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart