AMBH2D700368 Ambeed, Inc.

2,4-Bis((trimethylsilyl)oxy)pyrimidine

  • MDL number MFCD00054760
  • Cas Number : 10457-14-4
  • Molecular Weight : 256.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 31-33°C
Purity 98
Storage temp. Sealed in dry,2-8°C
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 264.8°C at 760 mmHg
Product Link https://www.ambeed.com/products/10457-14-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/10457-14-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart