AMBH2D700451 Ambeed, Inc.

3-(2,4-Dimethyl-1H-pyrrol-3-yl)propanoic acid

  • MDL number MFCD02684431
  • Cas Number : 54474-50-9
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 332.8°C at 760 mmHg
Product Link https://www.ambeed.com/products/54474-50-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/54474-50-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart